missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human COPZ2 Full-length ORF (NP_057513.1, 1 a.a. - 210 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00051226-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16129792

  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a subunit of the coatomer protein complex, a seven-subunit complex that functions in the formation of COPI-type, non-clathrin-coated vesicles. COPI vesicles function in the retrograde Golgi-to-ER transport of dilysine-tagged proteins. This gene is similar to a related family member, and the two encoded proteins form distinct isotypes of the coatomer protein complex. [provided by RefSeq]

Sequence: MQRPEAWPRPHPGEGAAAAQAGGPAPPARAGEPSGLRLQEPSLYTIKAVFILDNDGRRLLAKYYDDTFPSMKEQMVFEKNVFNKTSRTESEIAFFGGMTIVYKNSIDLFLYVVGSSYENELMLMSVLTCLFESLNHMLRKNVEKRWLLENMDGAFLVLDEIVDGGVILESDPQQVIQKVNFRADDGGLTEQSVAQVLQSAKEQIKWSLLK
Specifica

Specifica

NP_057513.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
49.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC23008
COPZ2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
51226
COPZ2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MQRPEAWPRPHPGEGAAAAQAGGPAPPARAGEPSGLRLQEPSLYTIKAVFILDNDGRRLLAKYYDDTFPSMKEQMVFEKNVFNKTSRTESEIAFFGGMTIVYKNSIDLFLYVVGSSYENELMLMSVLTCLFESLNHMLRKNVEKRWLLENMDGAFLVLDEIVDGGVILESDPQQVIQKVNFRADDGGLTEQSVAQVLQSAKEQIKWSLLK
RUO
COPZ2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali