missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CPE Partial ORF (NP_001864.1, 211 a.a. - 325 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00001363-Q01.10ug

Dettagli aggiuntivi : Peso : 0.02000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16013835

  • 335.00€ / 10µg
Spedizione stimata: 07-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a carboxypeptidase that cleaves C-terminal amino acid residues and is involved in the biosynthesis of peptide hormones and neurotransmitters, including insulin. It is a peripheral membrane protein. The protein specifically binds regulated secretory pathway proteins, including prohormones, but not constitutively secreted proteins. Mutations in this gene are implicated in type II diabetes. [provided by RefSeq]

Sequence: LLKNMKKIVDQNTKLAPETKAVIHWIMDIPFVLSANLHGGDLVANYPYDETRSGSAHEYSSSPDDAIFQSLARAYSSFNPAMSDPNRPPCRKNDDDSSFVDGTTNGGAWYSVPGG
Specifica

Specifica

NP_001864.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
38.39kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LLKNMKKIVDQNTKLAPETKAVIHWIMDIPFVLSANLHGGDLVANYPYDETRSGSAHEYSSSPDDAIFQSLARAYSSFNPAMSDPNRPPCRKNDDDSSFVDGTTNGGAWYSVPGG
RUO
CPE
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
1363
CPE (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CPE
Wheat Germ (in vitro)
GST
Liquid
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali