Learn More
Abcam Human CTSD knockout A-431 cell line
Codice prodotto. 32154977 Sfoglia Tutto Abcam Prodotti
Cambia vista
Click to view available options
Quantità:
1,000,000 Cells
2 x 1,000,000 Cells
Dimensione della confezione:
Pezzo
pezzi 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
32154977 1,000,000 Cells Pezzo
32140398 2 x 1,000,000 Cells pezzi 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 32154977 Fornitore Abcam N. del fornitore ab2618911000000Cells_
 Articolo soggetto a restrizioni
Spedizione stimata: 16-11-2026
Aggiungi al carrello
Aggiungi al carrello
missing translation for 'validMainframeMsg'
CTSD KO cell line available to order. KO validated by Next Generation Sequencing, Western blot. Free of charge wild type control available. Knockout achieved by CRISPR/Cas9 X = 22 bp deletion Frameshift = 100%. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. T...
CTSD KO cell line available to order. KO validated by Next Generation Sequencing, Western blot. Free of charge wild type control available. Knockout achieved by CRISPR/Cas9 X = 22 bp deletion Frameshift = 100%. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifica
Quantità 1,000,000 Cells
Status giuridico RUO
Titolo del prodotto
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.