missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human CXXC6 Partial ORF (NP_085128.1, 2038 a.a. - 2136 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00080312-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Sequence: RNHPTRLSLVFYQHKNLNKPQHGFELNKIKFEAKEAKNKKMKASEQKDQAANEGPEQSSEVNELNQIPSHKALTLTHDNVVTVSPYALTHVAGPYNHWVSpecifica
NP_085128.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RNHPTRLSLVFYQHKNLNKPQHGFELNKIKFEAKEAKNKKMKASEQKDQAANEGPEQSSEVNELNQIPSHKALTLTHDNVVTVSPYALTHVAGPYNHWV | |
RUO | |
TET1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
80312 | |
CXXC6 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CXXC6/FLJ10839/FLJ41442/KIAA1676/LCX/bA119F7.1 | |
TET1 | |
Recombinant | |
wheat germ expression system |