missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CYP26C1 Partial ORF (NP_899230.2, 423 a.a. - 522 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00340665-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16132827

  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme is involved in the catabolism of all-trans- and 9-cis-retinoic acid, and thus contributes to the regulation of retinoic acid levels in cells and tissues. This gene is adjacent to a related gene on chromosome 10q23.33. [provided by RefSeq]

Sequence: SPPEGFDPERFGAAREDSRGASSRFHYIPFGGGARSCLGQELAQAVLQLLAVELVRTARWELATPAFPAMQTVPIVHPVDGLRLFFHPLTPSVAGNGLCL
Specifica

Specifica

NP_899230.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SPPEGFDPERFGAAREDSRGASSRFHYIPFGGGARSCLGQELAQAVLQLLAVELVRTARWELATPAFPAMQTVPIVHPVDGLRLFFHPLTPSVAGNGLCL
RUO
CYP26C1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
340665
CYP26C1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ45301
CYP26C1
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali