Learn More
Abnova™ Human DAB2IP Partial ORF (NP_115941.1, 551 a.a. - 650 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00153090-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
DAB2IP is a Ras (MIM 190020) GTPase-activating protein (GAP) that acts as a tumor suppressor gene and is inactivated by methylation in prostate and breast cancers (Yano et al., 2005 [PubMed 15386433]).[supplied by OMIM]
Sequence: KSLSMVDLQDARTLDGEAGSPAGPDVLPTDGQAAAAQLVAGWPARATPVNLAGLATVRRAGQTPTTPGTSEGAPGRPQLLAPLSFQNPVYQMAAGLPLSPSpecifica
NP_115941.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KSLSMVDLQDARTLDGEAGSPAGPDVLPTDGQAAAAQLVAGWPARATPVNLAGLATVRRAGQTPTTPGTSEGAPGRPQLLAPLSFQNPVYQMAAGLPLSP | |
RUO | |
DAB2IP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
153090 | |
DAB2IP (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AF9Q34/AIP1/DIP1/2/FLJ39072/KIAA1743 | |
DAB2IP | |
Recombinant | |
wheat germ expression system |