missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DDB2 Partial ORF (NP_000098.1, 332 a.a. - 427 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00001643-Q02.10ug

Dettagli aggiuntivi : Peso : 0.02000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16053885

  • 310.00€ / 10µg


Spedizione stimata
14-01-2021
Aggiungi al carrello

Descrizione

Descrizione

Sequence: RHFQHLTPIKAAWHPRYNLIVVGRYPDPNFKSCTPYELRTIDVFDGNSGKMMCQLYDPESSGISSLNEFNPMGDTLASAMGYHILIWSQEEARTRK
Specifica

Specifica

NP_000098.1
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.3kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RHFQHLTPIKAAWHPRYNLIVVGRYPDPNFKSCTPYELRTIDVFDGNSGKMMCQLYDPESSGISSLNEFNPMGDTLASAMGYHILIWSQEEARTRK
RUO
DDB2
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
1643
DDB2 (Human) Recombinant Protein (Q02)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DDBB|FLJ34321|UV-DDB2
DDB2
Yes
Videos
SDS
Documenti