missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DEFB109 Full-length ORF (AAI48782.1, 1 a.a. - 87 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00641517-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16160814

  • 458.00€ / 25µg


Spedizione stimata
27-11-2020
Aggiungi al carrello

Descrizione

Descrizione

Sequence: MRLHLLLLILLLFSILLSPVRGGLGPAEGHCLNLFGVCRTDVCNIVEDQIGACRRRMKCCRAWWILMPIPTPLIMSDYQEPLKPNLK
Specifica

Specifica

AAI48782.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.52kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DEFB109
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
641517
DEFB109 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MRLHLLLLILLLFSILLSPVRGGLGPAEGHCLNLFGVCRTDVCNIVEDQIGACRRRMKCCRAWWILMPIPTPLIMSDYQEPLKPNLK
RUO
DEFB109
Yes
Videos
SDS
Documenti