Learn More
Abcam Human DGCR8 knockout HCT116 cell line
Codice prodotto. 32153013 Sfoglia Tutto Abcam Prodotti
Codice prodotto. 32153013 Fornitore Abcam N. del fornitore ab2873662x1000000Cells_
 Articolo soggetto a restrizioni
Spedizione stimata: 09-11-2026
Aggiungi al carrello
Aggiungi al carrello
missing translation for 'validMainframeMsg'
DGCR8 KO cell line available to order. KO validated by Next Generation Sequencing, Western blot. Free of charge wild type control available. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only knockout cells: select 1000000 Cells/vial.
DGCR8 KO cell line available to order. KO validated by Next Generation Sequencing, Western blot. Free of charge wild type control available. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement