Learn More
Abcam Human DICER1 knockout HEK293T cell line [4-25]
Descrizione
DICER1 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Expression plasmids encoding a pair of TALEN proteins designed to cleave the human dicer (hdcr) gene at nucleotide (nt) 1095 of the hDcr cDNA sequence, in coding exon 11. Clone 4-25. Knockout. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifica
Specifica
| Quantità | 2 x 1,000,000 Cells |
| Status giuridico | RUO |