Learn More
Abnova™ Human DPM1 Partial ORF (NP_003850, 161 a.a. - 260 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00008813-Q01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Dolichol-phosphate mannose (Dol-P-Man) serves as a donor of mannosyl residues on the lumenal side of the endoplasmic reticulum (ER). Lack of Dol-P-Man results in defective surface expression of GPI-anchored proteins. Dol-P-Man is synthesized from GDP-mannose and dolichol-phosphate on the cytosolic side of the ER by the enzyme dolichyl-phosphate mannosyltransferase. Human DPM1 lacks a carboxy-terminal transmembrane domain and signal sequence and is regulated by DPM2. [provided by RefSeq]
Sequence: RKIISRGANFLTQILLRPGASDLTGSFRLYRKEVLEKLIEKCVSKGYVFQMEMIVRARQLNYTIGEVPISFVDRVYGESKLGGNEIVSFLKGLLTLFATTSpecifica
NP_003850 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RKIISRGANFLTQILLRPGASDLTGSFRLYRKEVLEKLIEKCVSKGYVFQMEMIVRARQLNYTIGEVPISFVDRVYGESKLGGNEIVSFLKGLLTLFATT | |
RUO | |
DPM1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8813 | |
DPM1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDGIE/MPDS | |
DPM1 | |
Recombinant | |
wheat germ expression system |