Learn More
Abcam Human EGFR knockout A549 cell line
Codice prodotto. 32157373 Sfoglia Tutto Abcam Prodotti
Cambia vista
Click to view available options
Quantità:
1,000,000 Cells
2 x 1,000,000 Cells
Dimensione della confezione:
Pezzo
pezzi 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
32157373 1,000,000 Cells Pezzo
32140550 2 x 1,000,000 Cells pezzi 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 32157373 Fornitore Abcam N. del fornitore ab2863941000000Cells_
 Articolo soggetto a restrizioni
Spedizione stimata: 23-11-2026
Aggiungi al carrello
Aggiungi al carrello
missing translation for 'validMainframeMsg'
EGFR KO cell line available to order. KO validated by Next Generation Sequencing. Free of charge wild type control available. 6 bp (allele 1) 3 bp (allele 2) and 10 bp deletion after amino acid Ser160 of the WT EGFR protein. To order both knockout and wild-type control cells: select 2 x 1000000 ...
EGFR KO cell line available to order. KO validated by Next Generation Sequencing. Free of charge wild type control available. 6 bp (allele 1) 3 bp (allele 2) and 10 bp deletion after amino acid Ser160 of the WT EGFR protein. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifica
Quantità 1,000,000 Cells