missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human EMR4 Partial ORF (XP_377506, 21 a.a. - 93 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00326342-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16132787

  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene is a member of the EGF-TM7 receptor gene family which is thought to play a role in leukocyte adhesion and migration. In other vertebrates, including nonhuman primates, this gene encodes a protein containing N-terminal EGF domains and a C-terminal transmembrane domain. Sequence evidence for the human gene, however, indicates nucleotide deletion in the genomic sequence would result in frameshift and early termination of translation. A protein expressed by this gene would be soluble rather than expressed on the cell surface. As the encoded protein has not been detected, this gene may represent a transcribed pseudogene. [provided by RefSeq]

Sequence: GSEAKNSGASCPPCPKYASCHNSTHCTCEDGFRARSGRTYFHDSSEKCEDINECETGLAKCKYKAYCRNKVGG
Specifica

Specifica

XP_377506
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.77kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GSEAKNSGASCPPCPKYASCHNSTHCTCEDGFRARSGRTYFHDSSEKCEDINECETGLAKCKYKAYCRNKVGG
RUO
EMR4P
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
326342
EMR4 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EMR4/FIRE/GPR127/PGR16
EMR4P
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali