Learn More
Abcam Human ETS2 knockout HeLa cell line
Codice prodotto. 32155433 Sfoglia Tutto Abcam Prodotti
Cambia vista
Click to view available options
Quantità:
1,000,000 Cells
2 x 1,000,000 Cells
Dimensione della confezione:
Pezzo
pezzi 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
32155433 1,000,000 Cells Pezzo
32142622 2 x 1,000,000 Cells pezzi 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 32155433 Fornitore Abcam N. del fornitore ab2749451000000Cells_
 Articolo soggetto a restrizioni
Spedizione stimata: 27-11-2026
TERT KO cell line available to order. KO validated by Next Generation Sequencing. Free of charge wild type control available. 63 bp deletion (allele 1) and 1 bp insertion after Lys168 (allele 2) of the WT protein, Knockout. To order both knockout and wild-type control cells: select 2 x 1000000 C...
TERT KO cell line available to order. KO validated by Next Generation Sequencing. Free of charge wild type control available. 63 bp deletion (allele 1) and 1 bp insertion after Lys168 (allele 2) of the WT protein, Knockout. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifica
Quantità 1,000,000 Cells
Status giuridico RUO