Learn More
Abcam Human ETV7 knockout A549 cell line
Codice prodotto. 32142656
Cambia vista
Click to view available options
2x1000000 Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto.
32142656 2x1000000 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 32142656 Fornitore Abcam N. del fornitore ab2670752x1000000Cells_
 Articolo soggetto a restrizioni
Spedizione stimata: 13-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
ETV7 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 13 bp deletion in exon 5 and 19 bp deletion in exon 5. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only knockou...
ETV7 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 13 bp deletion in exon 5 and 19 bp deletion in exon 5. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantità 2 x 1,000,000 Cells
Status giuridico RUO
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.