missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FABP2 Partial ORF (NP_000125.1, 1 a.a. - 90 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00002169-Q01.25ug

Dettagli aggiuntivi : Peso : 0.02000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16063975

  • 458.00€ / 25µg


Spedizione stimata
22-10-2020
Aggiungi al carrello

Descrizione

Descrizione

Sequence: MAFDSTWKVDRSENYDKFMEKMGVNIVKRKLAAHDNLKLTITQEGNKFTVKESSAFRNIEVVFELGVTFNYNLADGTELRGTWSLEGNKL
Specifica

Specifica

NP_000125.1
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.64kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MAFDSTWKVDRSENYDKFMEKMGVNIVKRKLAAHDNLKLTITQEGNKFTVKESSAFRNIEVVFELGVTFNYNLADGTELRGTWSLEGNKL
RUO
FABP2
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
2169
FABP2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FABPI|I-FABP|MGC133132
FABP2
Yes
Videos
Documenti