missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human FGF R2 (IIIb) protein, His Tag (MALS verified)

453.47€ - 3329.77€

Specifica

Numero di accesso NP_075259.4
Peso molecolare 24.8 kDa
Sorgente Human FGFR2 (IIIb), His Tag (FGB-H5223) is expressed from human 293 cells (HEK293). It contains AA Pro 154 - Leu 358 (Accession # P21802-3).
Immunogeno Pro 154 - Leu 358
Requisiti di stoccaggio -20°C
Visualizza altre specifiche

Prodotti 2
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
17286200
Vedere i documenti Dettagli della promozione
ACROBiosystems
FGB-H5223-100UG
100 μg
453.47€
100µg
Visualizzare prodotti alternativi
Risparmia fino a 
»
Spedizione stimata: 11-04-2024
per visualizzare lo stock disponibile
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
17296200
Vedere i documenti Dettagli della promozione
ACROBiosystems
FGB-H5223-1MG
1 mg
3329.77€
1mg
Visualizzare prodotti alternativi
Risparmia fino a 
»
Spedizione stimata: 11-04-2024
per visualizzare lo stock disponibile
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
Descrizione

Descrizione

Tyrosine-protein kinase that acts as cell-surface receptor for fibroblast growth factors and plays an essential role in the regulation of cell proliferation, differentiation, migration and apoptosis, and in the regulation of embryonic development. Required for normal embryonic patterning, trophoblast function, limb bud development, lung morphogenesis, osteogenesis and skin development. Plays an essential role in the regulation of osteoblast differentiation, proliferation and apoptosis, and is required for normal skeleton development. Promotes cell proliferation in keratinocytes and immature osteoblasts, but promotes apoptosis in differentiated osteoblasts. Phosphorylates PLCG1, FRS2 and PAK4. Ligand binding leads to the activation of several signaling cascades. Activation of PLCG1 leads to the production of the cellular signaling molecules diacylglycerol and inositol 1,4,5-trisphosphate. Phosphorylation of FRS2 triggers recruitment of GRB2, GAB1, PIK3R1 and SOS1, and mediates activation of RAS, MAPK1/ERK2, MAPK3/ERK1 and the MAP kinase signaling pathway, as well as of the AKT1 signaling pathway. FGFR2 signaling is down-regulated by ubiquitination, internalization and degradation. Mutations that lead to constitutive kinase activation or impair normal FGFR2 maturation, internalization and degradation lead to aberrant signaling. Over-expressed FGFR2 promotes activation of STAT1.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.8 kDa. The protein migrates as 38-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifica

Specifica

NP_075259.4
Human FGFR2 (IIIb), His Tag (FGB-H5223) is expressed from human 293 cells (HEK293). It contains AA Pro 154 - Leu 358 (Accession # P21802-3).
-20°C
FGF R2 (IIIb)
Human
PBS, pH7.4
HEK293
Bio-Markers & CD Antigens
24.8 kDa
Pro 154 - Leu 358
FGF R2 (IIIb),FGFR2B,FGFR2
Unconjugated
His Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE.
Videos
SDS
Documenti

Documenti

Product Certifications