Learn More
Abcam Human FLAD1 knockout HEK-293T cell line
Codice prodotto. 32141160 Sfoglia Tutto Abcam Prodotti
Cambia vista
Click to view available options
Quantità:
2 x 1,000,000 Cells
Dimensione della confezione:
pezzi 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
32141160 2 x 1,000,000 Cells pezzi 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 32141160 Fornitore Abcam N. del fornitore ab2666992x1000000Cells_
 Articolo soggetto a restrizioni
Spedizione stimata: 30-11-2026
FLAD1 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 1 bp insertion in exon 3. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only knockout cells: select ...
FLAD1 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 1 bp insertion in exon 3. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifica
Quantità 2 x 1,000,000 Cells
Status giuridico RUO