missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FLJ45273 Partial ORF (NP_940863, 1 a.a. - 108 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00164832-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Sequence: MKNYEQALQDASAACQNEPLLIKGHQVKAQALSGLGRSKEVLKEFLYCLALNPECNSVKKEAQKVMCEVLFSATANVHENLTSSIQSRLKAQGHSHMNAQALLEEGDASpecifica
NP_940863 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKNYEQALQDASAACQNEPLLIKGHQVKAQALSGLGRSKEVLKEFLYCLALNPECNSVKKEAQKVMCEVLFSATANVHENLTSSIQSRLKAQGHSHMNAQALLEEGDA | |
RUO | |
LONRF2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
164832 | |
FLJ45273 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ45273/MGC126711/MGC126713/RNF192 | |
LONRF2 | |
Recombinant | |
wheat germ expression system |