Learn More
R&D Systems™ Human GABA-B R2 (P28476) VersaClone cDNA
Descrizione
The Human GABA-B R2 (P28476) VersaClone cDNA clone contains the complete ORF for GABA-B R2, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The GABA-B R2 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifica
Specifica
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Linea di prodotti | R&D Systems |
| Quantità | 10 μg |
For Reasearch Use Only; Not for Use in Humans