Learn More
Human Gas6 Antibody, R&D Systems™
GREENER_CHOICE
Codice prodotto. 18702527
Cambia vista
Click to view available options
Quantità:
100 μg
25 μg
Dimensione della confezione:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18702527 25 μg 25µg
18772147 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 18702527 Fornitore R&D Systems N. del fornitore MAB8852SP
Mouse Monoclonal Antibody
Gas6 Monoclonal specifically detects Gas6 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifica
Antigene Gas6
Applicazioni ELISA
Classificazione Monoclonal
Clone 100140
Coniugato Unconjugated
Diluizione ELISA
Formulazione Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
N. accesso geni NP_000811
Alias gene AXLLG, AXLLGAXL stimulatory factor, AXSFAXL receptor tyrosine kinase ligand, DKFZp666G247, FLJ34709, GAS-6, growth arrest-specific 6, growth arrest-specific protein 6
Specie ospite Mouse
Immunogeno Mouse myeloma cell line NS0-derived recombinant human Gas6, Asp118-Ala678, Accession No. NP_000811
Metodo di purificazione Protein A or G purified from cell culture supernatant
Quantità 25 μg
Primario o secondario Primary
ID gene (immissione) 2621.0
Test di specificità Detects human Gas6 in direct ELISAs.
Ricostituzione Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Forma Supernatant
Isotype IgG1