missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GATA3 Partial ORF (NP_002042.1, 169 a.a. - 248 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00002625-Q01.10ug
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Sequence: TPGSAGSARQDEKECLKYQVPLPDSMKLESSHSRGSMTALGGASSSTHHPITTYPPYVPEYSSGLFPPSSLLGGSPTGFGSpecifica
NP_002042.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.54kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
TPGSAGSARQDEKECLKYQVPLPDSMKLESSHSRGSMTALGGASSSTHHPITTYPPYVPEYSSGLFPPSSLLGGSPTGFG | |
RUO | |
GATA3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2625 | |
GATA3 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HDR/MGC2346/MGC5199/MGC5445 | |
GATA3 | |
Recombinant | |
wheat germ expression system |