Learn More
Abnova™ Human GNB1L Full-length ORF (NP_443730.1, 1 a.a. - 327 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00054584-P01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a G-protein beta-subunit-like polypeptide which is a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This protein contains 6 WD repeats and is highly expressed in the heart. The gene maps to the region on chromosome 22q11, which is deleted in DiGeorge syndrome, trisomic in derivative 22 syndrome and tetrasomic in cat-eye syndrome. Therefore, this gene may contribute to the etiology of those disorders. Transcripts from this gene share exons with some transcripts from the C22orf29 gene. [provided by RefSeq]
Sequence: MTAPCPPPPPDPQFVLRGTQSPVHALHFCEGAQAQGRPLLFSGSQSGLVHIWSLQTRRAVTTLDGHGGQCVTWLQTLPQGRQLLSQGRDLKLCLWDLAEGRSAVVDSVCLESVGFCRSSILAGGQPRWTLAVPGRGSDEVQILEMPSKTSVCALKPKADAKLGMPMCLRLWQADCSSRPLLLAGYEDGSVVLWDVSEQKVCSRIACHEEPVMDLDFDSQKARGISGSAGKALAVWSLDWQQALQVRGTHELTNPGIAEVTIRPDRKILATAGWDHRIRVFHWRTMQPLAVLAFHSAAVQCVAFTADGLLAAGSKDQRISLWSLYPRASpecifica
NP_443730.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
62kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DGCRK3/FKSG1/GY2/KIAA1645/WDR14/WDVCF | |
GNB1L | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
54584 | |
GNB1L (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTAPCPPPPPDPQFVLRGTQSPVHALHFCEGAQAQGRPLLFSGSQSGLVHIWSLQTRRAVTTLDGHGGQCVTWLQTLPQGRQLLSQGRDLKLCLWDLAEGRSAVVDSVCLESVGFCRSSILAGGQPRWTLAVPGRGSDEVQILEMPSKTSVCALKPKADAKLGMPMCLRLWQADCSSRPLLLAGYEDGSVVLWDVSEQKVCSRIACHEEPVMDLDFDSQKARGISGSAGKALAVWSLDWQQALQVRGTHELTNPGIAEVTIRPDRKILATAGWDHRIRVFHWRTMQPLAVLAFHSAAVQCVAFTADGLLAAGSKDQRISLWSLYPRA | |
RUO | |
GNB1L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |