Descrizione
GNG4 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp insertion in exon 3 and 2 bp deletion in exon 3 and 4 bp deletion in exon 3. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifica
Specifica
| Quantità | 2 x 1,000,000 Cells |
| Status giuridico | RUO |