missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HGF Partial ORF (NP_000592, 619 a.a. - 728 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00003082-Q02.25ug

Dettagli aggiuntivi : Peso : 0.02000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16064115

  • 458.00€ / 25µg


Spedizione stimata
19-10-2020
Aggiungi al carrello

Descrizione

Descrizione

Sequence: YTGLINYDGLLRVAHLYIMGNEKCSQHHRGKVTLNESEICAGAEKIGSGPCEGDYGGPLVCEQHKMRMVLGVIVPGRGCAIPNRPGIFVRVAYYAKWIHKIILTYKVPQS
Specifica

Specifica

NP_000592
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
YTGLINYDGLLRVAHLYIMGNEKCSQHHRGKVTLNESEICAGAEKIGSGPCEGDYGGPLVCEQHKMRMVLGVIVPGRGCAIPNRPGIFVRVAYYAKWIHKIILTYKVPQS
RUO
HGF
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
3082
HGF (Human) Recombinant Protein (Q02)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
F-TCF|HGFB|HPTA|SF
HGF
Yes
Videos
Documenti