Learn More
R&D Systems™ Human HLA-G (NP_002118) VersaClone cDNA
Descrizione
The Human HLA-G (NP_002118) VersaClone cDNA clone contains the complete ORF for HLA G, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The HLA G insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Specifica
Specifica
| Gene | HLA G |
| Numero di accesso | NP_002118 |
| Concentrazione | 10 μg at 0.2 μg/μL |
| Formulazione | cDNA is provided in 10 mM Tris-Cl |
| Tipo di inserto | 1030bp |
| pH | 8.5 |
| ID gene (immissione) | 3135 |
| Alias gene | b2 microglobulin, HLA class I histocompatibility antigen, alpha chain G, HLA class I molecule, HLA G, HLA G antigen, HLA-6.0, HLAG, HLA-G histocompatibility antigen, class I, G, major histocompatibility complex, class I, G, MHC Class I Antigen G, MHC-G, sHLA-G |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Specie | Human |
| Vedi altri risultati |