missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HSD11B2 Full-length ORF (AAH64536.1, 1 a.a. - 405 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

dietro preventivo

Specifica

Nome HSD11B2 (Human) Recombinant Protein (P01)
Recombinant Yes
Metodo di purificazione Glutathione Sepharose 4 Fast Flow
Peso molecolare 70.5kDa
Requisiti di stoccaggio Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Visualizza altre specifiche

Descrizione

Descrizione

Sequence: MERWPWPSGGAWLLVAARALLQLLRSDLRLGRPLLAALALLAALDWLCQRLLPPPAALAVLAAAGWIALSRLARPQRLPVATRAVLITGCDSGFGKETAKKLDSMGFTVLATVLELNSPGAIELRTCCSPRLRLLQMDLTKPGDISRVLEFTKAHTTSTGLWGLVNNAGHNEVVADAELSPVATFRSCMEVNFFGALELTKGLLPLLRSSRGRIVTVGSPAGDMPYPCLGAYGTSKAAVALLMDTFSCELLPWGVKVSIIQPGCFKTESVRNVGQWEKRKQLLLANLPQELLQAYGKDYIEHLHGQFLHSLRLAMSDLTPVVDAITDALLAARPRRRYYPGQGLGLMYFTHYYLPEGLRRRFLQAFFISHCLPRALQPGQPGTTPPQDAAQDPNLSPGPSPAVAR
Specifica

Specifica

HSD11B2 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
70.5kDa
Videos
SDS
Documenti

Documenti

Product Certifications