Learn More
Abcam Human ICOSLG knockout A549 cell line
Codice prodotto. 32143013 Sfoglia Tutto Abcam Prodotti
Codice prodotto. 32143013 Fornitore Abcam N. del fornitore ab2890551000000Cells_
 Articolo soggetto a restrizioni
Spedizione stimata: 27-10-2026
Aggiungi al carrello
Aggiungi al carrello
missing translation for 'validMainframeMsg'
ICOSLG KO cell line available to order. KO validated by Next Generation Sequencing. Free of charge wild type control available. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only knockout cells: select 1000000 Cells/vial.
ICOSLG KO cell line available to order. KO validated by Next Generation Sequencing. Free of charge wild type control available. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement