missing translation for 'onlineSavingsMsg'
Learn More

Human IFN-beta Antibody, R&D Systems™
GREENER_CHOICE

Codice prodotto. p-7195026
Change view
Click to view available options
Quantità:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità unitSize
18752107 25 μg 25µg
18723047 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18752107 Supplier R&D Systems Supplier No. MAB8145SP

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IFN-beta Monoclonal specifically detects IFN-beta in Human samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigene IFN-beta
Applicazioni Immunohistochemistry
Classificazione Monoclonal
Clone 1020344
Coniugato Unconjugated
Diluizione Immunohistochemistry 5-25 ug/mL
Formulazione Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
N. accesso geni P01574
Alias gene Fibroblast interferon, IFB, IFBIFNB, IFF, IFNB, IFNB1, IFNbeta, IFN-beta, interferon beta, interferon, beta 1, fibroblast, MGC96956
Specie ospite Mouse
Immunogeno Chinese Hamster Ovary cell line CHO-derived human IFN-beta , Met22-Asn187, Accession # P01574
Metodo di purificazione Protein A or G purified from hybridoma culture supernatant
Quantità 25 μg
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 3456
Ricostituzione Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Forma Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.