missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IGF-II/IGF2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB2921-SP
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IGF-II/IGF2 Monoclonal specifically detects IGF-II/IGF2 in Human samples. It is validated for Western Blot.
Specifica
| IGF-II | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C11orf43, chromosome 11 open reading frame 43, FLJ22066, FLJ44734, GRDF, IGF2, IGF-2, IGFII, IGF-II, insulin-like growth factor 2 (somatomedin A), insulin-like growth factor II, insulin-like growth factor type 2, MSA, PEG2, PP9974, somatomedin-A | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3481 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 75014 | |
| Western Blot 2 ug/mL | |
| P01344.1 | |
| IGF2 | |
| E. coli-derived recombinant human IGF-II Ala25-Glu91 Accession # P01344.1 | |
| 25 μg | |
| Primary | |
| Detects human IGF-II in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows approximately 10 - 50% cross-reactivity with recombinant mouse IGF-II. | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto