Learn More
ACROBiosystems Human IL-15 Protein, His Tag
MALS verified
522.57€ - 4062.99€
Specifica
Peso molecolare | 14.8 kDa |
---|---|
Sorgente | Human IL-15, His Tag (IL5-H52H8) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 (Accession # P40933-1). |
Immunogeno | Asn 49 - Ser 162 |
Requisiti di stoccaggio | -20°C |
Alias gene | IL-15,Interleukin-15,MGC9721 |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
17196411
|
ACROBiosystems
IL5-H52H8-1MG |
1 mg |
4062.99€
1mg |
Spedizione stimata: 30-04-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
17106421
|
ACROBiosystems
IL5-H52H8-50UG |
50 μg |
522.57€
50µg |
Spedizione stimata: 30-04-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
Interleukin 15 is also known as IL15, IL-15, and is a cytokine with structural similarity to IL-2. Like IL-2, IL-15 binds to and signals through the IL-2/IL-15 beta chain (CD122) and the common gamma chain (gamma-C, CD132). IL-15 is secreted by mononuclear phagocytes (and some other cells) following infection by virus(es). This cytokine induces cell proliferation of natural killer cells; cells of the innate immune system whose principal role is to kill virally infected cells. Interleukin 15 (IL-15) regulates T and natural killer (NK) cell activation and proliferation. Survival signals that maintain memory T cells in the absence of antigen are provided by IL-15. This cytokine is also implicated in NK cell development. In rodent lymphocytes, IL-15 prevents apoptosis by inducing an apoptosis inhibitor, BCL2L1/BCL-x(L). IL-15 has been shown to enhance the anti-tumor immunity of CD8+ T cells in pre-clinical models. A phase I clinical trial to evaluate the safety, dosing, and anti-tumor efficacy of IL-15 in patients with metastatic melanoma and renal cell carcinoma (kidney cancer) has begun to enroll patients at the National Institutes of Health.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 14.8 kDa. The protein migrates as 17-25 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.Specifica
14.8 kDa | |
Asn 49 - Ser 162 | |
IL-15,Interleukin-15,MGC9721 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
Human IL-15, His Tag (IL5-H52H8) is expressed from human 293 cells (HEK293). It contains AA Asn 49 - Ser 162 (Accession # P40933-1). | |
-20°C | |
IL-15 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Cytokines and Growth Factors |