missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-18 ProQuantum Immunoassay Kit

Codice prodotto. 17154163
Cambia vista
Click to view available options
Quantità:
96 Tests
Dimensione della confezione:
pezzi 96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
17154163 96 Tests pezzi 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17154163 Fornitore Invitrogen™ N. del fornitore A35613

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

ELISA

Utilizing proximity-based amplification technology, the assay combines the analyte specificity of high-affinity antibody-antigen binding with the signal detection and amplification capabilities of real-time PCR to achieve a simple yet powerful next-generation protein quantitation platform. A user-friendly workflow combined with intuitive software for analytics enables sample-to-answer in just 2 hours.

  • High sensitivity-detect low levels of protein with greater sensitivity than traditional methods
  • Broad dynamic range-≥5 logarithmic units, minimizing sample dilutions to ensure they fall within the range
  • Small sample consumption-use 2–5 μL of sample (compared to 75 μL for triplicate wells with other methods)
  • Fast, easy workflow-2 hours from sample to answer, with no wash steps
  • No proprietary instrument to purchase-runs on any real-time PCR instrument

The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein can specifically interact with this cytokine, and thus negatively regulate its biological activity.
TRUSTED_SUSTAINABILITY

Specifica

Numero di accesso cytokine; iboctadekin; IFN-gamma-inducing factor; IGIF; il 18; IL-1 gamma; IL18; IL-18; IL1F4; IL-1 g; ILN; interferon gamma inducing factor; interferon gamma-inducing factor; interferon-gamma-inducing factor; Interleukin; interleukin 18; interleukin 18 (interferon-gamma-inducing factor); interleukin-1 gamma; Interleukin18; Interleukin-18; interleukin-18 proprotein; interleukin-18-like protein; MGC12320
Intervallo saggio 0.064 to 1000 pg/mL
Assay Sensitivity <0.05 pg/mL
Coniugato FAM™
Product Type ProQuantum
Tipo di campione Plasma, Serum, Supernatant
Da utilizzare con (apparecchiatura) qPCR Instruments
ID gene (immissione) 3606
Simbolo del gene IL18
Contenuto kit Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase
Quantità 96 Tests
Status giuridico RUO
Volume del campione Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL
Requisiti di stoccaggio -20°C
Bersaglio IL-18
Durata del test 30 mins
Tempo totale saggio 2 hrs
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.