Learn More
Abnova™ Human INF2 Full-length ORF (NP_116103.1, 1 a.a. - 234 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifica
Numero di accesso | NP_116103.1 |
---|---|
Da utilizzare con (applicazione) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulazione | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
ID gene (immissione) | 64423 |
Peso molecolare | 52.4kDa |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
16173613
|
Abnova™
H00064423-P02.25UG |
25 ug |
508.00€
25µg |
Spedizione stimata: 04-06-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
16163613
|
Abnova™
H00064423-P02.10UG |
10 ug |
335.00€
10µg |
Spedizione stimata: 04-06-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
Actin filaments grow only when actin monomers have access to the fast-growing barbed end of the filament. The geometry of the filament network depends on the actions of the ARP2/3 complex (MIM 604221) and members of the formin family, such as INF2. The ARP2/3 complex binds to the sides of preexisting filaments and nucleates filaments whose barbed ends are quickly blocked by capping proteins, producing brush-like structures, such as those found at the leading edges of crawling cells. In contrast, formins bind to the barbed ends of growing filaments and protect them from capping, creating long filaments that can be cross-linked into bundles, such as those found in actin cables of yeast. Interaction of formins with actin barbed ends occurs through the formin homology-2 (FH2) domain. FH2 domains accelerate filament nucleation, move with the barbed end as the filament grows, and block capping of the barbed end by proteins such as gelsolin (GSN; MIM 137350). The FH1 domain of formins binds to profilin (see MIM 176610) and accelerates elongation from the FH2-bound barbed ends (Bindschadler and McGrath, 2004 [PubMed 15466701]; Chhabra and Higgs, 2006 [PubMed 16818491]).[supplied by OMIM]
Sequence: MSVKEGAQRKWAALKEKLGPQDSDPTEANLESADPELCIRLLQMPSVVNYSGLRKRLEGSDGGWMVQFLEQSGLDLLLEALARLSGRGVARISDALLQLTCVSCVRAVMNSRQGIEYILSNQGYVRQLSQALDTSNVMVKKQVFELLAALCIYSPEGHVLTLDALDHYKTVCSQQYRFSIVMNELSGSDNVPYVVTLLSVINAVILGPEDLRARTQLRNEFIGLQLLDVLARLRSpecifica
NP_116103.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
52.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSVKEGAQRKWAALKEKLGPQDSDPTEANLESADPELCIRLLQMPSVVNYSGLRKRLEGSDGGWMVQFLEQSGLDLLLEALARLSGRGVARISDALLQLTCVSCVRAVMNSRQGIEYILSNQGYVRQLSQALDTSNVMVKKQVFELLAALCIYSPEGHVLTLDALDHYKTVCSQQYRFSIVMNELSGSDNVPYVVTLLSVINAVILGPEDLRARTQLRNEFIGLQLLDVLARLR | |
RUO | |
INF2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
64423 | |
INF2 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C14orf151/C14orf173/DKFZp762A0214/FLJ22056/MGC13251/pp9484 | |
INF2 | |
Recombinant | |
wheat germ expression system |