missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KAL1 Partial ORF (NP_000207, 548 a.a. - 657 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00003730-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16127071

  • 349.00€ / 10µg
Spedizione stimata: 02-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Mutations in this gene cause the X-linked Kallmann syndrome. The encoded protein is similar in sequence to proteins known to function in neural cell adhesion and axonal migration. In addition, this cell surface protein is N-glycosylated and may have anti-protease activity. [provided by RefSeq]

Sequence: LAKPENLSASFIVQDVNITGHFSWKMAKANLYQPMTGFQVTWAEVTTESRQNSLPNSIISQSQILPSDHYVLTVPNLRPSTLYRLEVQVLTPGGEGPATIKTFRTPELPP
Specifica

Specifica

NP_000207
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LAKPENLSASFIVQDVNITGHFSWKMAKANLYQPMTGFQVTWAEVTTESRQNSLPNSIISQSQILPSDHYVLTVPNLRPSTLYRLEVQVLTPGGEGPATIKTFRTPELPP
RUO
KAL1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
3730
KAL1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ADMLX/HHA/KAL/KALIG-1/KMS
KAL1
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali