Learn More
Abnova™ Human KCNA4 Partial ORF (NP_002224, 562 a.a. - 652 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00003739-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes - shaker, shaw, shab, and shal - have been identified in Drosophila, and each has been shown to have human homolog(s). This gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily. This member contains six membrane-spanning domains with a shaker-type repeat in the fourth segment. It belongs to the A-type potassium current class, the members of which may be important in the regulation of the fast repolarizing phase of action potentials in heart and thus may influnce the duration of cardiac action potential. The coding region of this gene is intronless, and the gene is clustered with genes KCNA3 and KCNA10 on chromosome 1. [provided by RefSeq]
Sequence: NFNYFYHRETENEEQTQLTQNAVSCPYLPSNLLKKFRSSTSSSLGDKSEYLEMEEGVKESLCAKEEKCQGKGDDSETDKNNCSNAKAVETDSpecifica
NP_002224 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.75kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NFNYFYHRETENEEQTQLTQNAVSCPYLPSNLLKKFRSSTSSSLGDKSEYLEMEEGVKESLCAKEEKCQGKGDDSETDKNNCSNAKAVETD | |
RUO | |
KCNA4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3739 | |
KCNA4 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HBK4/HK1/HPCN2/HUKII/KCNA4L/KCNA8/KV1.4/PCN2 | |
KCNA4 | |
Recombinant | |
wheat germ expression system |