missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KCNAB3 Full-length ORF (NP_004723.2, 1 a.a. - 404 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00009196-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16173382

  • 349.00€ / 10µg
Spedizione stimata: 03-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. Four sequence-related potassium channel genes - shaker, shaw, shab, and shal - have been identified in Drosophila, and each has been shown to have human homolog(s). This gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily. This member is one of the beta subunits, which are auxiliary proteins associating with functional Kv-alpha subunits. This member and the KCNA5 gene product assemble into a heteromultimeric A-type channel that inactivates completely and is significantly faster than other A-type Kv channels. [provided by RefSeq]

Sequence: MQVSIACTEQNLRSRSSEDRLCGPRPGPGGGNGGPAGGGHGNPPGGGGSGPKARAALVPRPPAPAGALRESTGRGTGMKYRNLGKSGLRVSCLGLGTWVTFGSQISDETAEDVLTVAYEHGVNLFDTAEVYAAGKAERTLGNILKSKGWRRSSYVITTKIFWGGQAETERGLSRKHIIEGLRGSLERLQLGYVDIVFANRSDPNCPMEEIVRAMTYVINQGLALYWGTSRWGAAEIMEAYSMARQFNLIPPVCEQAEHHLFQREKVEMQLPELYHKIGVGSVTWYPLACGLITSKYDGRVPDTCRASIKGYQWLKDKVQSEDGKKQQAKVMDLLPVAHQLGCTVAQLAIAWCLRSEGVSSVLLGVSSAEQLIEHLGALQVLSQLTPQTVMEIDGLLGNKPHSKK
Specifica

Specifica

NP_004723.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
70.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AKR6A9/KCNA3.1B/KCNA3B/KV-BETA-3/MGC116886
KCNAB3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
9196
KCNAB3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
MQVSIACTEQNLRSRSSEDRLCGPRPGPGGGNGGPAGGGHGNPPGGGGSGPKARAALVPRPPAPAGALRESTGRGTGMKYRNLGKSGLRVSCLGLGTWVTFGSQISDETAEDVLTVAYEHGVNLFDTAEVYAAGKAERTLGNILKSKGWRRSSYVITTKIFWGGQAETERGLSRKHIIEGLRGSLERLQLGYVDIVFANRSDPNCPMEEIVRAMTYVINQGLALYWGTSRWGAAEIMEAYSMARQFNLIPPVCEQAEHHLFQREKVEMQLPELYHKIGVGSVTWYPLACGLITSKYDGRVPDTCRASIKGYQWLKDKVQSEDGKKQQAKVMDLLPVAHQLGCTVAQLAIAWCLRSEGVSSVLLGVSSAEQLIEHLGALQVLSQLTPQTVMEIDGLLGNKPHSKK
RUO
KCNAB3
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali