Learn More
R&D Systems™ Human KLHL6 (NP_569713.2) VersaClone cDNA
Specifica
Specifica
| Gene | KLHL6 |
| ID gene (immissione) | 89857 |
| Alias gene | FLJ00029, kelch-like 6 (Drosophila), kelch-like protein 6, kelch-like protein KLHL6 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantità | 10 μg |