Learn More
Abcam Human KMT2A knockout A549 cell line
Codice prodotto. 32142790
Codice prodotto. 32142790 Fornitore Abcam N. del fornitore ab2866751000000Cells_
 Articolo soggetto a restrizioni
Spedizione stimata: 05-10-2026
KMT2A KO cell line available to order. KO validated by Next Generation Sequencing. Free of charge wild type control available. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only knockout cells: select 1000000 Cells/vial.
KMT2A KO cell line available to order. KO validated by Next Generation Sequencing. Free of charge wild type control available. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement