Learn More
Abcam Human LAMTOR2 knockout HeLa cell line
Codice prodotto. 32139427 Sfoglia Tutto Abcam Prodotti
Cambia vista
Click to view available options
Quantità:
2 x 1,000,000 Cells
Dimensione della confezione:
pezzi 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
32139427 2 x 1,000,000 Cells pezzi 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 32139427 Fornitore Abcam N. del fornitore ab2658282x1000000Cells_
 Articolo soggetto a restrizioni
Spedizione stimata: 16-11-2026
Aggiungi al carrello
Aggiungi al carrello
missing translation for 'validMainframeMsg'
LAMTOR2 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp deletion in exon 2 and 7 bp deletion in exon 2. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To o...
LAMTOR2 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp deletion in exon 2 and 7 bp deletion in exon 2. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifica
Quantità 2 x 1,000,000 Cells