Learn More
Human LAP (TGF-beta 1) Antibody, R&D Systems™
Codice prodotto. 18161911
Cambia vista
Click to view available options
Quantità:
25 μg
500 μg
Dimensione della confezione:
25µg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18161911 500 μg 500µg
18779638 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 18161911 Fornitore R&D Systems N. del fornitore MAB2463
Mouse Monoclonal Antibody has been used in 8 publications
LAP (TGF-beta 1) Monoclonal specifically detects LAP (TGF-beta 1) in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifica
Antigene LAP (TGF-beta 1)
Applicazioni Flow Cytometry, CyTOF
Classificazione Monoclonal
Clone 27232
Coniugato Unconjugated
Diluizione Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulazione Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N. accesso geni P01137
Alias gene CED, DPD1, LAP, LAP (TGFbeta 1), TGFB, TGFB1, TGFbeta, transforming growth factor beta 1
Simboli geni TGFB1
Specie ospite Mouse
Immunogeno S. frugiperda insect ovarian cell line Sf 21-derived recombinant human LAP (TGF-beta 1) Leu30-Arg278 (Cys33Ser) Accession # P01137
Metodo di purificazione Protein A or G purified from hybridoma culture supernatant
Quantità 500 μg
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 7040
Test di specificità Detects human LAP (TGF-beta 1) in direct ELISAs. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) TGF-beta 1, rhTGF-beta 2, rhTGF-beta 1.2, rhTGF-beta 3, or rhTGF-alpha .
Ricostituzione Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Forma Purified
Isotype IgG1
For Research Use Only