missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LBX2 Partial ORF (NP_001009812.1, 95 a.a. - 194 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00085474-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16161037

  • 302.00€ / 10µg


Spedizione stimata
19-11-2020
Aggiungi al carrello

Descrizione

Descrizione

Sequence: ELERRFVFQKYLAPSERDGLATRLGLANAQVVTWFQNRRAKLKRDVEEMRADVASLRALSPEVLCSLALPEGAPDPGLCLGPAGPDSRPHLSDEEIQVDD
Specifica

Specifica

NP_001009812.1
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
ELERRFVFQKYLAPSERDGLATRLGLANAQVVTWFQNRRAKLKRDVEEMRADVASLRALSPEVLCSLALPEGAPDPGLCLGPAGPDSRPHLSDEEIQVDD
RUO
LBX2
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
85474
LBX2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LP3727
LBX2
Yes
Videos
Documenti