missing translation for 'onlineSavingsMsg'
Learn More

Human LILRA5/CD85f Antibody, R&D Systems™
GREENER_CHOICE

Codice prodotto. 18179030
Cambia vista
Click to view available options
Quantità:
100 μg
Dimensione della confezione:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18179030 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 18179030 Fornitore R&D Systems N. del fornitore MAB67541

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Mouse Monoclonal Antibody

ILT11/LILRA5 Monoclonal specifically detects ILT11/LILRA5 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifica

Antigene ILT11/LILRA5
Applicazioni Western Blot
Classificazione Monoclonal
Clone 711827
Concentrazione LYOPH
Coniugato Unconjugated
Diluizione Western Blot 2 ug/mL
Formulazione Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
N. accesso geni A6NI73
Alias gene CD85, CD85 antigen-like family member F, CD85F, CD85f antigen, ILT11, ILT-11, ILT11CD85f, Immunoglobulin-like transcript 11, leukocyte Ig-like receptor 9, Leukocyte immunoglobulin-like receptor 9, leukocyte immunoglobulin-like receptor subfamily A member 5, leukocyte immunoglobulin-like receptor subfamily A member 5 soluble, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 5, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LILRB7, LIR9, LIR-9, LIR9immunoglobulin-like transcript 11 protein, member 7
Simboli geni LILRA5
Specie ospite Mouse
Immunogeno Chinese hamster ovary cell line CHO-derived recombinant human LILRA5/CD85f Gly42-Arg268 Accession # A6NI73
Metodo di purificazione Protein A or G purified from hybridoma culture supernatant
Quantità 100 μg
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 353514
Test di specificità Detects human LILRA5/CD85f in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse LILRC1, recombinant rat LILRC1, recombinant human (rh) ILT7/LILRA4, rhILT4, rhILT5, or rhLILRB4/LIR5 is observed. In Western blots, no cross-reactivity with rhILT7/LILRA4 or rhILT5 is observed.
Ricostituzione Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Forma Purified
Isotype IgG2a
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.