missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human LRRC15 (NP_570843) VersaClone cDNA
Descrizione
The Human LRRC15 (NP_570843) VersaClone cDNA clone contains the complete ORF for LRRC15, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The LRRC15 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature
Specifica
Specifica
| Product Type | cDNA |
| Gene | hLRRC15 |
| Numero di accesso | NP_570843 |
| Concentrazione | 10 μg at 0.2 μg/μL |
| Formulazione | cDNA is provided in 10mM Tris-Cl, pH 8.5 |
| Tipo di inserto | 1759bp |
| ID gene (immissione) | 131578 (Human), 74488 (Mouse), 246296 (Rat) |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Specie | Human |
| Quantità | 10 μg |
Titolo del prodotto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?