Learn More
Abcam Human MAGEA6 knockout HeLa cell line
Codice prodotto. 32156172 Sfoglia Tutto Abcam Prodotti
Cambia vista
Click to view available options
2x1000000 Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto.
32156172 2x1000000 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 32156172 Fornitore Abcam N. del fornitore ab2652972x1000000Cells_
 Articolo soggetto a restrizioni
Spedizione stimata: 26-10-2026
MAGEA6 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp deletion in exon 3 and Insertion of the selection cassette in exon 3. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial....
MAGEA6 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp deletion in exon 3 and Insertion of the selection cassette in exon 3. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifica
Quantità 2 x 1,000,000 Cells
Status giuridico RUO