Learn More
Abcam Human MAP3K11 (MLK3) knockout A549 cell line
Codice prodotto. 32154085
Cambia vista
Click to view available options
Quantità:
1,000,000 Cells
2 x 1,000,000 Cells
Dimensione della confezione:
Pezzo
pezzi 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
32154085 1,000,000 Cells Pezzo
32139762 2 x 1,000,000 Cells pezzi 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 32154085 Fornitore Abcam N. del fornitore ab2671691000000Cells_
 Articolo soggetto a restrizioni
Spedizione stimata: 19-10-2026
Aggiungi al carrello
Aggiungi al carrello
missing translation for 'validMainframeMsg'
MAP3K11 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp deletion in exon 5 and 1 bp insertion in exon 5 and 288 bp insertion in exon 5. To order both knockout and wild-type control cells: selec...
MAP3K11 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp deletion in exon 5 and 1 bp insertion in exon 5 and 288 bp insertion in exon 5. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifica
Quantità 1,000,000 Cells