missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MAST4 Partial ORF (XP_291141, 2605 a.a. - 2699 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00375449-Q01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Sequence: LPLESHHPDPNTMGGASHRDRALSVTATVGETKGKDPAPAQPPPARKQNVGRDVTKPSPAPNTDRPISLSNEKDFVVRQRRGKESLRSSPHKKALSpecifica
XP_291141 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.19kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LPLESHHPDPNTMGGASHRDRALSVTATVGETKGKDPAPAQPPPARKQNVGRDVTKPSPAPNTDRPISLSNEKDFVVRQRRGKESLRSSPHKKAL | |
RUO | |
MAST4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
375449 | |
MAST4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686E18148/DKFZp686N1467/FLJ16540/FLJ33039/KIAA0303 | |
MAST4 | |
Recombinant | |
wheat germ expression system |