missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MCCC1 Full-length ORF (AAH04214.1, 1 a.a. - 725 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00056922-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16132343

  • 528.00€ / 25µg
Spedizione stimata: 02-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes the large subunit of 3-methylcrotonyl-CoA carboxylase. This enzyme functions as a heterodimer and catalyzes the carboxylation of 3-methylcrotonyl-CoA to form 3-methylglutaconyl-CoA. Mutations in this gene are associated with 3-Methylcrotonylglycinuria, an autosomal recessive disorder of leucine catabolism. [provided by RefSeq]

Sequence: MAAASAVSVLLVAAERNRWHRLPSLLLPPRTWVWRQRTMKYTTATGRNITKVLIANRGEIACRVMRTAKKLGVQTVAVYSEADRNSMHVDMADEAYSIGPAPSQQSYLSMEKIIQVAKTSAAQAIHPGCGFLSENMEFAELCKQEGIIFIGPPPSAIRDMGIKSTSKSIMAAAGVPVVEGYHGEDQSDQCLKEHARRIGYPVMIKAVRGGGGKGMRIVRSEQEFQEQLESARREAKKSFNDDAMLIEKFVDTPRHVEVQVFGDHHGNAVYLFERDCSVQRRHQKIIEEAPAPGIKSEVRKKLGEAAVRAAKAVNYVGAGTVEFIMDSKHNFCFMEMNTRLQVEHPVTEMITGTDLVEWQLRIAAGEKIPLSQEEITLQGHAFEARIYAEDPSNNFMPVAGPLVHLSTPRADPSTRIETGVRQGDEVSVHYDPMIAKLVVWAADRQAALTKLRYSLRQYNIVGLPTNIDFLLNLSGHPEFEAGNVHTDFIPQHHKQLLLSRKAAAKESLCQAALGLILKEKAMTDTFTLQAHDQFSPFSSSSGRRLNISYTRNMTLKDGKNNVAIAVTYNHDGSYSMQIEDKTFQVLGNLYSEGDCTYLKCSVNGVASKAKLIILENTIYLFSKEGSIEIDIPVPKYLSSVSSQETQGGPLAPMTGTIEKVFVKAGDKVKAGDSLMVMIAMKMEHTIKSPKDGTVKKVFYREGAQANRHTPLVEFEEEESDKRESE
Specifica

Specifica

AAH04214.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
106.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686B20267/FLJ25545/MCC-B/MCCA
MCCC1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
56922
MCCC1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAASAVSVLLVAAERNRWHRLPSLLLPPRTWVWRQRTMKYTTATGRNITKVLIANRGEIACRVMRTAKKLGVQTVAVYSEADRNSMHVDMADEAYSIGPAPSQQSYLSMEKIIQVAKTSAAQAIHPGCGFLSENMEFAELCKQEGIIFIGPPPSAIRDMGIKSTSKSIMAAAGVPVVEGYHGEDQSDQCLKEHARRIGYPVMIKAVRGGGGKGMRIVRSEQEFQEQLESARREAKKSFNDDAMLIEKFVDTPRHVEVQVFGDHHGNAVYLFERDCSVQRRHQKIIEEAPAPGIKSEVRKKLGEAAVRAAKAVNYVGAGTVEFIMDSKHNFCFMEMNTRLQVEHPVTEMITGTDLVEWQLRIAAGEKIPLSQEEITLQGHAFEARIYAEDPSNNFMPVAGPLVHLSTPRADPSTRIETGVRQGDEVSVHYDPMIAKLVVWAADRQAALTKLRYSLRQYNIVGLPTNIDFLLNLSGHPEFEAGNVHTDFIPQHHKQLLLSRKAAAKESLCQAALGLILKEKAMTDTFTLQAHDQFSPFSSSSGRRLNISYTRNMTLKDGKNNVAIAVTYNHDGSYSMQIEDKTFQVLGNLYSEGDCTYLKCSVNGVASKAKLIILENTIYLFSKEGSIEIDIPVPKYLSSVSSQETQGGPLAPMTGTIEKVFVKAGDKVKAGDSLMVMIAMKMEHTIKSPKDGTVKKVFYREGAQANRHTPLVEFEEEESDKRESE
RUO
MCCC1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali