Learn More
Abcam Human MELK knockout HCT116 cell line
Product Code. 32141598 Shop All Abcam Products
Change view
Click to view available options
Quantità:
2 x 1,000,000 Cells
Unit Size:
pezzi 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità Unit Size
32141598 2 x 1,000,000 Cells pezzi 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32141598 Supplier Abcam Supplier No. ab2668962x1000000Cells_
 Articolo soggetto a restrizioni
Spedizione stimata: 07-12-2026
MELK KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp deletion in exon 9 and Insertion of the selection cassette in exon 9. To order both knockout and wild-type control cells: select 2 x 100000...
MELK KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp deletion in exon 9 and Insertion of the selection cassette in exon 9. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantità 2 x 1,000,000 Cells
Status giuridico RUO