Learn More
R&D Systems™ Human mGluR7 (NP_000835) VersaClone cDNA
Codice prodotto. 18750648
Cambia vista
Click to view available options
Quantità:
10 μg
Dimensione della confezione:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18750648 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18750648 Fornitore R&D Systems™ N. del fornitore RDC1001
Please to purchase this item. Need a web account? Register with us today!
Glutamate receptor, metabotropic 7
The Human mGluR7 (NP_000835) VersaClone cDNA clone contains the complete ORF for mGluR7, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The mGluR7 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Specifica
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantità 10 μg
For Reasearch Use Only; Not for Use in Humans