missing translation for 'onlineSavingsMsg'
Learn More

Human MORF4L2 (MRGX) knockout HCT116 cell lysate, 1 Kit

Codice prodotto. 32142218 Sfoglia Tutto Abcam Prodotti
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 32142218 Fornitore Abcam N. del fornitore ab2585211Kit
 Articolo soggetto a restrizioni
Spedizione stimata: 23-11-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Human MORF4L2 (MRGX) knockout HCT116 cell/tissue lysate.

  • Species: Human
  • Tissue: MORF4L2 (MRGX) knockout HCT116
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.